THREADED SPACER FOR 25/30 MM ID 90CM LEN 1 * 1 items
Catalog # MAISAS2018-025000
Supplier: DR MAISCH
THREADED SPACER FOR 25/30 MM ID 90CM LEN 1 * 1 items
Catalog # MAISAS2018-025000
Supplier: DR MAISCH
Frequently Bought Together
Accessories for UV lamps and cabinets
Supplier: CAMAG
Avantor® ACE® C18-PFP, HPLC Columns, 5 µm
Supplier: Avantor
VWR®, Safety Waste Caps
Supplier: VWR Collection
Supplies for Agilent Instruments 7700 Series ICP-MS
Supplier: Agilent
UHPLC columns, ReproSil®-Pur C18-aQ
Supplier: DR MAISCH
Screw caps with 8 mm septa
Supplier: Agilent
Screw caps, 9 mm, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
HPLC columns, Hypersil GOLD™ SAX
Supplier: Thermo Fisher Scientific
Fused silica tubing
Supplier: Agilent
ISAspher HPLC Columns, Diol, HILIC
Supplier: ISERA
HPLC columns, ZORBAX® 80 Å StableBond®
Supplier: Agilent
TSKgel® gel filtration columns, PW-type
Supplier: TOSOH Bioscience
GC septa, three layer
Supplier: HAMILTON BONADUZ
Microvolume syringes 10 µl
Supplier: Trajan Scientific and Medical
Coated polysaccharide columns, normal phase, CHIRALPAK®
Supplier: CHIRAL TECHNOLOGIES
Avantor® ACE® AQ, HPLC Columns, 3 µm
Supplier: Avantor
LC vials and closures
Supplier: Agilent