Furnace ashing lvt9/11/b510 tMax1100 with liFt Door anD over-teMPerature liMiter anD therMocouPle hole in the rear wall 1 * 1 items
NABELV095H11NLTZ
Furnace ashing lvt9/11/b510 tMax1100 with liFt Door anD over-teMPerature liMiter anD therMocouPle hole in the rear wall 1 * 1 items
NABELV095H11NLTZ
Frequently Bought Together
Reaction Stations, RS-200 and RS-250
: Cole-Parmer
Modular Heating Blocks and Accessories for VWR® Block Heaters
: VWR Collection
Furnace rsv 50/250/11 w/controller b500 1 * 1 items
: NABERTHERM