Order Entry
Slovenia
ContactUsLinkComponent
Furnace MuFFle lt15/13/c550 tMax1300 with with liFt Door Door anD over-teMPerature liMiter anD therMocouPle hole in the rear wall 1 * 1 items
Catalog # NABEL-155H3ANOTZ
Supplier:  NABERTHERM
Furnace MuFFle lt15/13/c550 tMax1300 with with liFt Door Door anD over-teMPerature liMiter anD therMocouPle hole in the rear wall 1 * 1 items
Catalog # NABEL-155H3ANOTZ
Supplier:  NABERTHERM
Frequently Bought Together