Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ASEPTCOR® U-Series 16 PFA Schlauch Nennweite: 1,ID 25.40mm x AD 37.00mm 1 * 20 m
Catalog # MSPPACU.16.100.20
Supplier:
MARKET SOURCE PART PROCESS
Expand image
ASEPTCOR® U-Series 16 PFA Schlauch Nennweite: 1,ID 25.40mm x AD 37.00mm 1 * 20 m
Catalog # MSPPACU.16.100.20
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
VWR® Nitrile Extra Light, Nitrile Gloves
Supplier: VWR Collection
Neoprene gloves, Microflex® NeoTouch® 25-101 / 25-201
Supplier: Ansell
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Nitrile Light Long, Nitrile Gloves
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Boxes for Broken Glass
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™ Xtra™
Supplier: KIMBERLY CLARK
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR® Stockholm, Safety Spectacles
Supplier: VWR Collection
VWR®, Latex Gloves
Supplier: VWR Collection
VWR® Nitrile Green Nitrile gloves
Supplier: VWR Collection