Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66S3S00123
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66S3S00123
Supplier: Sartorius Balances
Frequently Bought Together
Accessories for orbital shakers PSU-10i
Supplier: GRANT INSTRUMENTS
Accessories for reciprocating shakers, HS 260
Supplier: IKA
REPLACEMENT STIRRER 1 * 1 items
Supplier: GERBER FUNKE
Grinding jar EasyFit for Planetary Ball Mills, tungsten carbide, 250ml 1 * 1 items
Supplier: Retsch GmbH
VWR®, Multi Purpose Clamps, 3-Prong
Supplier: VWR Collection
Mats for analogue/digital and reciprocating shakers
Supplier: OHAUS
Propeller head 50mmwide for SS stirrer range 1 * 1 items
Supplier: Cole Parmer
Accessories for Thermo CO₂ resistant orbital shaker
Supplier: Thermo Fisher Scientific
Overhead stirrer tools, anchor stirrers
Supplier: BOCHEM
Rigid threaded couplings, BOLA
Supplier: Bohlender
Distance sieve ZM 300 titanium trapezoid holes 2.00 mm 1 * 1 items
Supplier: Retsch GmbH
Accessories for magnetic stirrers, TWISTER
Supplier: IKA
Stainless steel sieves for rotor beater mills, SR 200 and SR 300
Supplier: Retsch GmbH
Overhead shakers, Reax 20 series
Supplier: Heidolph Instruments GmbH & Co.KG
Accessories for laboratory mixers
Supplier: WARING BLENDOR
Accessories for orbital shaker, VXR basic Vibrax®
Supplier: IKA