Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
Catalog # SARBMCA66S3CF99H
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
Catalog # SARBMCA66S3CF99H
Supplier: Sartorius Balances
Frequently Bought Together
Accessories for planetary ball mills, PM100, PM100 CM, PM200 and PM400
Supplier: Retsch GmbH
Rack - chrome plated 1 * 1 items
Supplier: Binder
Accessories for Memmert UN/UNplus and UF/UFplus ovens
Supplier: MEMMERT
Factory Fitted Options for Binder FP Heating Chambers
Supplier: Binder
Heating tapes, HST
Supplier: HORST
Precision™ General purpose water baths
Supplier: THERMO ELECTRON LED
Incubators, IN/INplus (natural convection)
Supplier: MEMMERT
Multi-position magnetic stirrers, Variomag Biosystem
Supplier: THERMO H + P LABORTECHNIK
Gas burner, powerjet/powerjet 2
Supplier: WARTEWIG
Dispersing elements for homogenisers
Supplier: IKA
Heating mantles, WHU2ER series, PILZ®
Supplier: WINKLER