Order Entry
Slovenia
ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
  SARBMCA66S3CF24M
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
  SARBMCA66S3CF24M
Frequently Bought Together