Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
Catalog # SARBMCA66S3CF14M
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
Catalog # SARBMCA66S3CF14M
Supplier: Sartorius Balances
Frequently Bought Together
VWR®, Tube Rotator
Supplier: VWR Collection
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
VWR® Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
Magnetic stirring bars
Supplier: VELP SCIENTIFIC
VWR® VS-CV, Magnetic Stirrers
Supplier: VWR Collection
Accessories for tube mill control
Supplier: IKA
MR Hei-Mix S Magnetic Stirrer
Supplier: Heidolph Instruments GmbH & Co.KG
Vortex mixer, VORTEX Genius 3
Supplier: IKA
VWR® lab dancer, Vortex Mixer
Supplier: VWR Collection
Magnetic stirrer, Mini MR standard IKAMAG®
Supplier: IKA
Accessories for Magnetic Hotplate Stirrers
Supplier: IKA
Findenser™ Air condenser for Magnetic Stirrer
Supplier: Heidolph Instruments GmbH & Co.KG
VWR® VMS-A, Magnetic Hotplate Stirrer
Supplier: VWR Collection
VWR®, Disposable Pestles, Microtubes and Cordless Motor for Pellet Mixing
Supplier: VWR Collection
Magnetic hotplate stirrer, RH basic 2 IKAMAG®
Supplier: IKA
Magnetic stirring bars sets, IKAFLON®
Supplier: IKA
Accessories for Retsch sieves
Supplier: Retsch GmbH
Overhead stirrer tools, dissolver stirrers
Supplier: IKA
Stainless steel sieves for ZM 200
Supplier: Retsch GmbH