Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66S3CF13
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66S3CF13
Supplier: Sartorius Balances
Frequently Bought Together
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
Magnetic hotplate stirrer, RCT basic
Supplier: IKA
VWR®, Mini vortex mixer
Supplier: VWR Collection
Magnetic hotplate stirrers, ARE/AREX 5 series
Supplier: VELP SCIENTIFIC
Vortex mixers, Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Magnetic stirrers, MST and MST Digital
Supplier: VELP SCIENTIFIC
Overhead stirrer tools, 4-blade stirrers
Supplier: IKA
Accessories for hotplates, VHP-C Series
Supplier: IKA
Vortex mixers, CLASSIC and WIZARD
Supplier: VELP SCIENTIFIC
VWR® VMS Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
Magnetic hotplate stirrers, C-MAG HS 7 control
Supplier: IKA
Hei-PLATE Mix 'n' Heat Core+ Magnetic Hotplate Stirrers
Supplier: Heidolph Instruments GmbH & Co.KG
Accessories for GRINDOMIX GM 200
Supplier: Retsch GmbH
Electronic overhead stirrers, Hei-TORQUE core
Supplier: Heidolph Instruments GmbH & Co.KG
Accessories for Retsch sieves
Supplier: Retsch GmbH
Stirrer Bearings, glass
Supplier: Bohlender
Accessories for DrySyn® Heating Blocks
Supplier: ASYNT