Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA66S3CE4I
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA66S3CE4I
Supplier: Sartorius Balances
Frequently Bought Together
Accessories for overhead stirrer
Supplier: IKA
Heating and drying ovens, Universal UF/UFplus (forced convection)
Supplier: MEMMERT
Shelves for Heratherm® incubators
Supplier: Thermo Fisher Scientific
Cole-Parmer® HM-200 Flask/Funnel Heating Mantles
Supplier: Cole-Parmer
Sample splitters, RT 6,5 - RT 75
Supplier: Retsch GmbH
with integrated power controller, temperature range 450C, 2000 W, power supply 230 V 1 * 1 items
Supplier: witeg Labortechnik
Cole-Parmer® SH-200 Series Shaker and SI-200 Series Shaking Incubator Accessories
Supplier: Cole Parmer
Accessories for mixer mills MM 200 and MM 400
Supplier: Retsch GmbH
Modular heating blocks
Supplier: OHAUS
Tubings, insulated
Supplier: LAUDA
Accessories for CR®, Viton® and reinforced tubings
Supplier: JULABO GmbH
Accessories for heating circulators
Supplier: JULABO GmbH
Accessories for TW series water baths
Supplier: JULABO GmbH
Cyclone with filter bag with collecting receptacle 3 litres 1 * 1 items
Supplier: Retsch GmbH
Factory fitted options for Memmert UNpa series Paraffin ovens
Supplier: MEMMERT