Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
Catalog # SARBMCA66S3CE13H
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
Catalog # SARBMCA66S3CE13H
Supplier: Sartorius Balances
Frequently Bought Together
Accessories for RZR overhead stirrers
Supplier: Heidolph Instruments GmbH & Co.KG
2 Viton sleeves 2 mm dia. 1 * 1 items
Supplier: JULABO GmbH
Accessories for Q500/Q700 Sonicators
Supplier: QSONICA LLC
Gas burner, powerjet/powerjet 2
Supplier: WARTEWIG
Bunsen burners, with needle valve, 1300 °C
Supplier: BOCHEM
Shaking incubator, digital, benchtop, ES-20
Supplier: GRANT INSTRUMENTS
Heating thermostats with stainless steel bath, ECO series
Supplier: LAUDA
Hose connections for Huber circulators
Supplier: HUBER
Accessories for JB Academy, JB Nova, SUB Aqua Pro, SUB Aqua Plus, JB Aqua Plus and SBB Aqua Plus baths
Supplier: GRANT INSTRUMENTS
Laboratory grinding mill, MultiDrive control
Supplier: IKA
Cole-Parmer® WB-300 Series General-Purpose Digital Water Baths, Stuart
Supplier: Cole Parmer
Temperature Probes for Cole-Parmer® CN-200 Series Digital Heating Controllers
Supplier: Cole-Parmer
Carrier for round bottom flasks, IKA®
Supplier: IKA
High temperature tube furnaces with Tmax 1600 °C, RHTC series
Supplier: NABERTHERM
Heating and drying ovens, Universal UN/UNplus (natural convection)
Supplier: MEMMERT
Accessories for vortex mixers Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Heating Mantles, WHG2, WHG2H, WHG2R and WHG4R
Supplier: WINKLER