Order Entry
Slovenia
ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 12 | 61 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA66P3S0023I
Supplier:  Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 12 | 61 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA66P3S0023I
Supplier:  Sartorius Balances
Frequently Bought Together