Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 12 | 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66P3S00123
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 12 | 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66P3S00123
Supplier: Sartorius Balances
Frequently Bought Together
Accessories for magnetic hotplate stirrers
Supplier: Heidolph Instruments GmbH & Co.KG
VWR®, Mini Vortex Mixers
Supplier: VWR Collection
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
VWR® VS-CV, Magnetic Stirrers
Supplier: VWR Collection
Vortex mixers, ZX4 and TX4
Supplier: VELP SCIENTIFIC
Programmable Vortex Mixers, Genie Series
Supplier: SCIENTIFIC INDUSTRIES ELMIS
VWR® Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
Magnetic hotplate stirrers, ARE/AREX 5 series
Supplier: VELP SCIENTIFIC
Magnetic hotplate stirrers, C-MAG HS 7 control
Supplier: IKA
Magnetic stirrers, TWISTER
Supplier: IKA
Electronic overhead stirrer, NANOSTAR 7.5 digital
Supplier: IKA
Overhead stirrer tools, 4-blade stirrers
Supplier: IKA
Accessories for tube mill control
Supplier: IKA
Magnetic stirrers, MST and MST Digital
Supplier: VELP SCIENTIFIC
MaxQ™ 8000 Incubating Shakers
Supplier: Thermo Fisher Scientific
Hei-PLATE Mix 'n' Heat Core+ Magnetic Hotplate Stirrers
Supplier: Heidolph Instruments GmbH & Co.KG