Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 12 | 61 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
Catalog # SARBMCA66P3CF12M
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 12 | 61 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
Catalog # SARBMCA66P3CF12M
Supplier: Sartorius Balances
Frequently Bought Together
Cole-Parmer® WBU-200 Series Unheated Water Baths
Supplier: Cole-Parmer
Incubators, IN/INplus (natural convection)
Supplier: MEMMERT
Water baths, unstirred, SUB Aqua Pro series
Supplier: GRANT INSTRUMENTS
Furnace MuFFle l5/14/b510 tMax1400 with FlaP Door 1 * 1 items
Supplier: NABERTHERM
[EN]MAGNETIC HOTPLATE-STIRRER CERAMIC 1 1 * 1 ST
Supplier: VWR Collection
Dri-Block Aluminum Heating Block Insert, 18 X 20 mm diameter 1 * 1 items
Supplier: Cole-Parmer
Accessories for Memmert UN/UNplus and UF/UFplus ovens
Supplier: MEMMERT
Heating Mantles, WHG2, WHG2H, WHG2R and WHG4R
Supplier: WINKLER
Factory Fitted Options for Binder FP Heating Chambers
Supplier: Binder