Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 12 | 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66P3CE4
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 12 | 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66P3CE4
Supplier: Sartorius Balances
Frequently Bought Together
Accessories for Memmert UN/UNplus and UF/UFplus ovens
Supplier: MEMMERT
Door gasket - made of FKM, silicone-free 1 * 1 items
Supplier: Binder
Planetary micro mill, PULVERISETTE 7 classic line
Supplier: FRITSCH
Cyclone with filter bag with collecting receptacle 3 litres 1 * 1 items
Supplier: Retsch GmbH
Block Heater Insert, Double Wide, 384 Well Plate 1 * 1 items
Supplier: Cole Parmer
SR test tube/microtube racks
Supplier: GRANT INSTRUMENTS
Mats for analogue/digital and reciprocating shakers
Supplier: OHAUS
Accessories for vortex mixers Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Accessories for Fuego SCS Safety Gas Burners
Supplier: WARTEWIG
Heating tapes, HBS
Supplier: HORST