Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 12 | 61 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
Catalog # SARBMCA66P3CE2M
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 12 | 61 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
Catalog # SARBMCA66P3CE2M
Supplier: Sartorius Balances
Frequently Bought Together
Disc 2 for capsules for autosampler 1 * 1 items
Supplier: VELP SCIENTIFIC
Vibratory Feeder DR 100 drive unit, 220-240 V, 60 Hz 1 * 1 items
Supplier: Retsch GmbH
MaxQ™ 8000 Incubating Shakers
Supplier: Thermo Fisher Scientific
StarFish multiple experiment workstation
Supplier: Heidolph Instruments GmbH & Co.KG
Distance sieve ZM 300 titanium trapezoid holes 2.00 mm 1 * 1 items
Supplier: Retsch GmbH
VWR®, Accessories for VWR® INCU-Line Shaking Incubators
Supplier: VWR Collection
Ultrasonic converter, HD 5000
Supplier: BANDELIN-ELECTRONIC
I-MAG BR Bracket for stand rod I-MAG 1 * 1 items
Supplier: IKA
Overhead stirrer tools, paddle stirrers
Supplier: BOCHEM
Heating circulators, MAGIO™ MS and MX series
Supplier: JULABO GmbH
ROTATOR DRY POWDER 1 * 1 items
Supplier: INSTRUMENTS FOR RESEARCH