Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36S3S00234
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36S3S00234
Supplier: Sartorius Balances
Frequently Bought Together
Accessories for MA series
Supplier: Sartorius Balances
Gas lighter, piezoelectric
Supplier: USBECK Laborgeräte
Overhead stirrer tools, 4-blade stirrers
Supplier: IKA
Stirrer bearings, PTFE
Supplier: Bohlender
VWR®, Mini vortex mixer
Supplier: VWR Collection
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
Magnetic hotplate stirrer, RCT basic
Supplier: IKA
Accessories for hotplates, VHP-C Series
Supplier: IKA
VWR® VMS-A, Magnetic Hotplate Stirrer
Supplier: VWR Collection
VWR® VMS Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
VWR® lab dancer, Vortex Mixer
Supplier: VWR Collection
Magnetic stirrer, Topolino
Supplier: IKA
Cartridge 220G for cartridge burner
Supplier: USBECK Laborgeräte
Magnetic stirrer, Mini MR standard IKAMAG®
Supplier: IKA
Vortex mixers, MS 3
Supplier: IKA
Magnetic stirrers, MICROSTIRRER
Supplier: VELP SCIENTIFIC
Vortex mixers, Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
VWR®, Disposable Pestles, Microtubes and Cordless Motor for Pellet Mixing
Supplier: VWR Collection