0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
SARBMCA36S3S0013M
:
Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
SARBMCA36S3S0013M
:
Sartorius Balances
Frequently Bought Together
External measuring instrument with sensors for daylight and uv-light for ich 1 * 1 items
: MEMMERT
Overhead stirrer tools, anchor stirrers
: BOCHEM
VWR®, Covers for VWR® Professional and Advanced Hotplate Stirrers
: VWR Collection
Accessories for Optima™ heated circulating baths
: GRANT INSTRUMENTS
Accessories for Unheated Water Baths, Cole-Parmer®, WBU-200 Series
: Cole-Parmer
Cabel EU for NEXOPART Sieving Machines 1 * 1 items
: VWR Collection
Boiling baths, SBB Aqua Plus series
: GRANT INSTRUMENTS
Tissue grinders, Tenbroeck type, KIMBLE®
: DWK Life Sciences
Wall brackets: Tubular frame for wall mounting for Memmert UN/UNplus and UF/UFplus ovens
: MEMMERT
Propeller stirrer with 4 blades, 18/10 steel, L=800mm, D=70mm, shaft diameter 10mm 1 * 1 items
: BOCHEM