Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA36S3S00124I
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA36S3S00124I
Supplier: Sartorius Balances
Frequently Bought Together
Magnetic hotplate stirrers, ARE/AREX 5 series
Supplier: VELP SCIENTIFIC
VWR®, Mini vortex mixer
Supplier: VWR Collection
VWR® VMS-A, Magnetic Hotplate Stirrer
Supplier: VWR Collection
VWR®, Tube Rotator
Supplier: VWR Collection
Accessories for tube mill control
Supplier: IKA
Vortex mixer, VORTEX Genius 3
Supplier: IKA
Magnetic stirrers, MST and MST Digital
Supplier: VELP SCIENTIFIC
VWR® VMS Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
VWR® VV3, Vortex Mixer
Supplier: VWR Collection
VWR® lab dancer, Vortex Mixer
Supplier: VWR Collection
Cartridge 220G for cartridge burner
Supplier: USBECK Laborgeräte
Magnetic stirrer, Mini MR standard IKAMAG®
Supplier: IKA
Analytical mill, A 11 basic
Supplier: IKA
Vortex mixers, Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Accessories and stirring tools for ES, PW, BS, DLS, LH and DLH stirrers
Supplier: VELP SCIENTIFIC
Vortex mixers, RX3 and ZX3
Supplier: VELP SCIENTIFIC
Accessories for VWR® Advanced and Professional Hotplates and Hotplate Stirrers
Supplier: VWR Collection
VWR®, Accessories for Analogue, Digital and Pulsing Vortex Shakers
Supplier: VWR Collection
Laboratory mixers
Supplier: WARING BLENDOR