Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA36S3CF34I
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA36S3CF34I
Supplier: Sartorius Balances
Frequently Bought Together
Magnetic hotplate stirrers, C-MAG HS digital series
Supplier: IKA
VWR® DRY-Line® Prime, Drying Ovens with Natural Convection
Supplier: VWR Collection
Gas cartridge, CP250
Supplier: CAMPING GAZ
Cartridge burner
Supplier: USBECK Laborgeräte
Cartridge 220G for cartridge burner
Supplier: USBECK Laborgeräte
Burners
Supplier: USBECK Laborgeräte
VWR® VMS Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
Hose connections for Huber circulators
Supplier: HUBER
VWR®, Trays for VLSB Shaking Baths
Supplier: VWR Collection
Heating and drying ovens, Universal UN/UNplus (natural convection)
Supplier: MEMMERT
Temperature control units
Supplier: JULABO GmbH
Dual height adjustable vessel clamp for Solaris Shaker 18”x18” Platforms 1 * 1 items
Supplier: Thermo Fisher Scientific
Interchangeable blocks for block heaters, QBA, QBD and QBH
Supplier: GRANT INSTRUMENTS
Tubing, chloroprene and viton
Supplier: JULABO GmbH
Bath fluids
Supplier: JULABO GmbH
Accessories for Lauda ECO range circulators
Supplier: LAUDA