Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36S3CF2
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36S3CF2
Supplier: Sartorius Balances
Frequently Bought Together
Accessories for vortex mixers Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Homogeniser, T 25 digital ULTRA-TURRAX®
Supplier: IKA
Vortex mixers, ZX4 and TX4
Supplier: VELP SCIENTIFIC
VWR®, Mini vortex mixer
Supplier: VWR Collection
Magnetic stirrers, MICROSTIRRER
Supplier: VELP SCIENTIFIC
VWR®, Advanced hotplates
Supplier: VWR Collection
VWR®, Disposable Pestles, Microtubes and Cordless Motor for Pellet Mixing
Supplier: VWR Collection
Vortex mixers, RX3 and ZX3
Supplier: VELP SCIENTIFIC
Oil bath attachment for IKAMAG® and IKATHERM®
Supplier: IKA
Bunsen burner, cartridge type
Supplier: USBECK Laborgeräte
VWR® lab dancer, Vortex Mixer
Supplier: VWR Collection
VWR® VV3, Vortex Mixer
Supplier: VWR Collection
Boss Head
Supplier: Heidolph Instruments GmbH & Co.KG
Accessories for PURA™ water baths
Supplier: JULABO GmbH
VWR® Sample Splitters
Supplier: VWR Collection
Overhead stirrers, electronic, OHS digital and advance
Supplier: VELP SCIENTIFIC
Grindomix GM200 1 * 1 items
Supplier: Retsch GmbH