Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36S3CF14
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36S3CF14
Supplier: Sartorius Balances
Frequently Bought Together
Ceran® hotplates, LHG
Supplier: GESTIGKEIT HARRY
Gas burners, Fuego SCS basic RF
Supplier: WARTEWIG
Bunsen burners, LABOGAZ 206
Supplier: CAMPING GAZ
Cartridge burner, high temperature
Supplier: USBECK Laborgeräte
VWR® VMS-A, Magnetic Hotplate Stirrer
Supplier: VWR Collection
Magnetic stirrers, MST and MST Digital
Supplier: VELP SCIENTIFIC
Accessories for vortex mixers Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Magnetic hotplate stirrer, RH basic 2 IKAMAG®
Supplier: IKA
Gas lighter, Fire Boy
Supplier: FRANKEA
Safety gas tubing
Supplier: USBECK Laborgeräte
Cartridge 220G for cartridge burner
Supplier: R L ENTERPRISES
VWR® Standard 1000 Orbital Shaker
Supplier: VWR Collection
Cartridge 220G for cartridge burner
Supplier: USBECK Laborgeräte
Magnetic stirrer, Mini MR standard IKAMAG®
Supplier: IKA
Stirrers, propeller, with four paddles
Supplier: BOCHEM
VWR®, Stainless Steel Base Trays
Supplier: VWR Collection
Cole-Parmer® ST-200 Series Large-Capacity Magnetic Stirrers
Supplier: Cole Parmer
Electronic overhead stirrers, Hei-TORQUE core
Supplier: Heidolph Instruments GmbH & Co.KG
Crucible tongs
Supplier: NABERTHERM
Accessories for Retsch sieves
Supplier: Retsch GmbH