Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36S3CE23
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36S3CE23
Supplier: Sartorius Balances
Frequently Bought Together
Accessories for shakers
Supplier: Heidolph Instruments GmbH & Co.KG
Accessories for VWR® Series Homogenisers
Supplier: VWR Collection
StarFish multiple experiment workstation
Supplier: Heidolph Instruments GmbH & Co.KG
1000ml square media bottle clamp for Solaris Shaker Platforms 1 * 1 items
Supplier: Thermo Fisher Scientific
VWR®, Accessories for Analogue, Digital and Pulsing Vortex Shakers
Supplier: VWR Collection
Rigid threaded couplings, BOLA
Supplier: Bohlender
Overhead stirrer tools, maxi propeller stirrer, BOLA
Supplier: Bohlender
Air Jet Sieve Machine eControl Lite, e200 LS
Supplier: VWR Collection
Accessories for Thermo CO₂ resistant orbital shaker
Supplier: Thermo Fisher Scientific
LID CLOSER FOR PULVERISETTE 14 1 * 1 items
Supplier: FRITSCH