Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
SARBMCA36S3CE14
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
SARBMCA36S3CE14
Frequently Bought Together
Gas cartridges, Campingaz®
: CAMPING GAZ
Vortex mixers, Vortex-Genie®
: SCIENTIFIC INDUSTRIES ELMIS
Vortex mixers, MS 3
: IKA
Magnetic stirrers, MICROSTIRRER
: VELP SCIENTIFIC
VWR® VMS-A, Magnetic Hotplate Stirrer
: VWR Collection
VWR®, Mini Vortex Mixers
: VWR Collection
Accessories for MA series
: Sartorius Balances
Vortex mixers, ZX4 and TX4
: VELP SCIENTIFIC
VWR®, Mini vortex mixer
: VWR Collection
Vortex mixer, VORTEX 2
: IKA
Magnetic stirrers, MST and MST Digital
: VELP SCIENTIFIC
VWR® VMS Advanced Magnetic Hotplate Stirrers
: VWR Collection
Cartridge 220G for cartridge burner
: USBECK Laborgeräte
VWR® VOS 40 digital & VOS 60 control, Electronic Overhead Stirrers
: VWR Collection