Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36P3CF234
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36P3CF234
Supplier: Sartorius Balances
Frequently Bought Together
Gas burner, powerjet/powerjet 2
Supplier: WARTEWIG
Ceran® hotplates, LHG
Supplier: GESTIGKEIT HARRY
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
Magnetic hotplate stirrer, RCT basic
Supplier: IKA
VWR® VWB2, Water Baths
Supplier: VWR Collection
Roller Bar Platforms for VWR® Standard and Advanced Shakers
Supplier: VWR Collection
VWR® VMS-A, Magnetic Hotplate Stirrer
Supplier: VWR Collection
Cartridge 220G for cartridge burner
Supplier: USBECK Laborgeräte
VWR® lab dancer, Vortex Mixer
Supplier: VWR Collection
VWR® VMS Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
VWR®, Mini vortex mixer
Supplier: VWR Collection
Bunsen burners, LABOGAZ 206
Supplier: CAMPING GAZ
Hotplates, C-MAG HP series
Supplier: IKA
Heating and drying ovens, Universal UN/UNplus (natural convection)
Supplier: MEMMERT
Accessories for muffle furnaces, L 1/11-L 40/12 and LE 1/11 - LE 14/11
Supplier: NABERTHERM
VWR®, Disposable Pestles, Microtubes and Cordless Motor for Pellet Mixing
Supplier: VWR Collection
Crucible tongs
Supplier: NABERTHERM