Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Draft Shiel d Motors 1 * 1 items
Catalog # SARBMCA36P3CF23M
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Draft Shiel d Motors 1 * 1 items
Catalog # SARBMCA36P3CF23M
Supplier: Sartorius Balances
Frequently Bought Together
Accessories for shakers, KS 260 and HS 260
Supplier: IKA
Bathbridge kpl K6, K6s 08 1 * 1 items
Supplier: HUBER
Water baths and circulators, stainless steel, Optima™
Supplier: GRANT INSTRUMENTS
Accessories for alcohol lamps
Supplier: GLASWARENFABRIK KARL HECHT
VWR®, Dispersing Aggregates
Supplier: VWR Collection
Glass Liner for 100 mL vessel 1 * 1 items
Supplier: ASYNT
Heating and drying ovens, Universal UF/UFplus (forced convection)
Supplier: MEMMERT
Stirrer bearings, PTFE
Supplier: Bohlender
Magnetic stirring bars, Komet
Supplier: THERMO H + P LABORTECHNIK
Accessories for heating circulators
Supplier: JULABO GmbH
Sieves for POLYMIX® system PX-MFC 90 D
Supplier: KINEMATICA
Compact incubator with forced convection, Heratherm®
Supplier: Thermo Fisher Scientific
Accessories for Unheated Water Baths, Cole-Parmer®, WBU-200 Series
Supplier: Cole-Parmer