Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36P3CE3
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36P3CE3
Supplier: Sartorius Balances
Frequently Bought Together
Heat-on blocks
Supplier: Heidolph Instruments GmbH & Co.KG
Vortex mixers, Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
VWR®, Tissue Grinders, Potter-Elvehjem Type
Supplier: VWR Collection
Magnetic hotplate stirrer, RH basic 2 IKAMAG®
Supplier: IKA
Magnetic hotplate stirrers, ARE/AREX 6-series
Supplier: VELP SCIENTIFIC
Vortex mixers, RX3 and ZX3
Supplier: VELP SCIENTIFIC
Vortex mixers, ZX4 and TX4
Supplier: VELP SCIENTIFIC
Analytical mill, A 11 basic
Supplier: IKA
Cartridge 220G for cartridge burner
Supplier: USBECK Laborgeräte
VWR® lab dancer, Vortex Mixer
Supplier: VWR Collection
VWR® VMS Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
Accessories for Magnetic Hotplate Stirrers
Supplier: IKA
VWR®, Mini vortex mixer
Supplier: VWR Collection
Magnetic hotplate stirrers, ARE/AREX 5 series
Supplier: VELP SCIENTIFIC
Electronic overhead stirrer, NANOSTAR 7.5 digital
Supplier: IKA
Overhead stirrer tools, 4-blade stirrers
Supplier: IKA
Magnetic stirrers, MST and MST Digital
Supplier: VELP SCIENTIFIC
Bunsen burner, cartridge type
Supplier: USBECK Laborgeräte
VWR®, Magnetic Stirrer, Lab Disc
Supplier: VWR Collection