Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.002 mg, max. load 111 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA116S3CE123I
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.002 mg, max. load 111 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA116S3CE123I
Supplier: Sartorius Balances
Frequently Bought Together
Magnetic hotplate stirrer, RCT basic
Supplier: IKA
Ceran® hotplates, LHG
Supplier: GESTIGKEIT HARRY
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
Accessories for tube mill control
Supplier: IKA
VWR®, Mini vortex mixer
Supplier: VWR Collection
VWR® VMS Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
Gas lighter
Supplier: USBECK Laborgeräte
Cartridge 220G for cartridge burner
Supplier: USBECK Laborgeräte
VWR®, Lids for VWB2 Baths
Supplier: VWR Collection
Heating and drying ovens, Universal UF/UFplus (forced convection)
Supplier: MEMMERT
Magnetic stirring bars sets, IKAFLON®
Supplier: IKA
Water baths, PURA™
Supplier: JULABO GmbH
Stainless steel sieves for ZM 200
Supplier: Retsch GmbH
Accessories for heating mantles, PILZ®
Supplier: WINKLER
Overhead stirrers, electronic, OHS digital and advance
Supplier: VELP SCIENTIFIC
Stainless steel grids for Memmert UNpa ovens
Supplier: MEMMERT
Accessories for overhead shakers, Reax 2
Supplier: Heidolph Instruments GmbH & Co.KG
Heating tapes, HST
Supplier: HORST
Digital dry block heaters
Supplier: IKA