Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Guard Column Holders
Guard Column Holders
Guard column holder 'stand-alone' for 10 mm cartridges, ¹/₁₆" (10-32) thread on both sides
Catalog # ISERAA13-000004
Supplier:   ISERA
Guard Column Holders
Catalog # ISERAA13-000004
Supplier:  ISERA
Guard Column Holders
Guard column holder 'stand-alone' for 10 mm cartridges, ¹/₁₆" (10-32) thread on both sides
Price
74,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Guard column holder 'stand-alone' for 10 mm cartridges, ¹/₁₆" (10-32) thread on both sides
  • Seq:
    04