Guard Column Holders
Guard column holder for direct coupling of 10 mm cartridges to ISERA analytical columns (2 - 4,6 mm Int.Ø)
Catalog # ISERAA13-000002
Supplier: ISERA
Jump to:
Product Details & Documents
Specifications
- Description:Guard column holder for direct coupling of 10 mm cartridges to ISERA analytical columns (2 - 4,6 mm Int.Ø)
- Seq:02