Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
POTASSIUM PHOSPHATE DIBASIC ANH. FCC 1 * 2,5 kg
Catalog # SPCMPO205-2.5KG
Supplier:
Spectrum Chemical
Expand image
POTASSIUM PHOSPHATE DIBASIC ANH. FCC 1 * 2,5 kg
Catalog # SPCMPO205-2.5KG
Supplier:
Spectrum Chemical
Frequently Bought Together
Ammonium sulphate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium hydrogen phthalate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium chloride 99.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals