L(-)XYLOSE 1 * 5 g
Catalog # HIMERM1488-5G
Supplier: HIMEDIA
Frequently Bought Together
Petroleum spirit, 60…95 °C, TECHNICAL
Supplier: VWR Chemicals
Isooctane (Reag. Ph. Eur.) for analysis, ACS 1 * 2,5 L
Supplier: PanReac AppliChem
Bromoform 96% stabilised
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99.5+% for analysis
Supplier: Thermo Fisher Scientific
Ethyl acetate 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Toluene, extra dry over Molecular Sieve 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol standard for the analysis of tobacco
Supplier: VWR Chemicals
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrahydrofuran 99.8% unstabilised for HPLC
Supplier: Thermo Fisher Scientific
Tetrachloroethylene ≥99%
Supplier: Thermo Fisher Scientific
Liquid for manometer with sloping tube
Supplier: VWR Chemicals