Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
12 components: Cu 200mg/l Fe 100mg/l Zn 100mg/l Al 100mg/l Cr 40 1 * 100 mL
Catalog # CMPLRM071196L1.2
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
12 components: Cu 200mg/l Fe 100mg/l Zn 100mg/l Al 100mg/l Cr 40 1 * 100 mL
Catalog # CMPLRM071196L1.2
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach