(S, R, S)-AHPC-PEG2-AMINE HYDROCHLORIDE 1 * 100 mg
Catalog # APOSBIPC1038-100MG
Supplier: Apollo Scientific
(S, R, S)-AHPC-PEG2-AMINE HYDROCHLORIDE 1 * 100 mg
Catalog # APOSBIPC1038-100MG
Supplier: Apollo Scientific
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Diethyl ether stabilised Ph. Eur.
Supplier: VWR Chemicals
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
1,4-Dioxane, extra dry over Molecular Sieve 99,5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
n-Heptane 99+% for HPLC
Supplier: Thermo Fisher Scientific
GLYCEROL PURISS., MEETS ANALYTICAL SPECI 1 * 1 L
Supplier: Merck
1-Butanol 99%, extra pure
Supplier: Thermo Fisher Scientific