1-(4-Bromophenyl)propan-1-one,98% 1 * 25 g
Catalog # AMBDA229708-25G
Supplier: AMBEED
Frequently Bought Together
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Petroleum spirit, 60…80 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Hexane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Isobutanol ≥99%
Supplier: Thermo Fisher Scientific
Iodomethane, Sigma-Aldrich®
Supplier: Merck
1-Butanol for HPLC
Supplier: Thermo Fisher Scientific
(±)-1,2-Propanediol ACS
Supplier: Thermo Fisher Scientific
Cyclohexane, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals