(2S,3R)-2-Amino-3-(tert-butoxy)butanoic acid,97% 1 * 5 g
Catalog # AMBDA194968-5G
Supplier: AMBEED
Frequently Bought Together
2,2,4-Trimethylpentane ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone, BAKER, J.T.Baker®
Supplier: Avantor
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanolamine ≥99%, GPR RECTAPUR® for liquid scintillation counting
Supplier: VWR Chemicals
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
FAM Benzine DIN 51635, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
Benzyl alcohol ≥99%
Supplier: Thermo Fisher Scientific
Acetone, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ammonia solution 35% in water for HPLC
Supplier: Thermo Fisher Scientific