Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
(1S,2S)-(-)-1,2-Diphenylethylenediamine,98% 1 * 25 g
Catalog # AMBDA375912-25G
Supplier:
AMBEED
Expand image
(1S,2S)-(-)-1,2-Diphenylethylenediamine,98% 1 * 25 g
Catalog # AMBDA375912-25G
Supplier:
AMBEED
Frequently Bought Together
Methanol ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
Tetrahydrofuran, anhydrous 99.9% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Water for analysis
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific