Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
tert-Butyl 3-formyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate,98% 1 * 0,1 g
Catalog # AMBDA997048-100MG
Supplier:
AMBEED
Expand image
tert-Butyl 3-formyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate,98% 1 * 0,1 g
Catalog # AMBDA997048-100MG
Supplier:
AMBEED
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
Diethyl ether stabilised, TECHNICAL, rectified
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99%, TECHNICAL
Supplier: VWR Chemicals
2,2,4-Trimethylpentane 99+% ACS
Supplier: Thermo Fisher Scientific
Diiodomethane 99+% stabilised
Supplier: Thermo Fisher Scientific
Water for LC-MS 1 * 2,5 L
Supplier: PanReac AppliChem