0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
tert-Butyl 6-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate,95% 1 * 0,1 g
AMBDA764388-100MG
:
AMBEED
tert-Butyl 6-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate,95% 1 * 0,1 g
AMBDA764388-100MG
:
AMBEED
Frequently Bought Together
Turpentine oil
: Roth Carl
Isooctane for UV, IR, HPLC, ACS 1 * 1 L
: PanReac AppliChem
3-(Chloromethyl)benzoyl chloride 98%
: Thermo Fisher Scientific
Acetone (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
: PanReac AppliChem
Ethyl acetate ≥99.5% for HPLC
: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
: Thermo Fisher Scientific
Chloroform 99+% stabilised, extra pure
: Thermo Fisher Scientific
Ethyl Acetate, 99.5% for synthesis 1 * 5 L
: PanReac AppliChem
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
Methyl ethyl ketone ACS
: Thermo Fisher Scientific
Toluene ≥99.7% for HPLC
: Thermo Fisher Scientific