4-((4-Methylpiperazin-1-yl)methyl)-3-(trifluoromethyl)aniline,98% 1 * 25 g
AMBDA568431-25G
: AMBEED
Frequently Bought Together
Hydrogen peroxide 35%, J.T.Baker®
: Avantor
1-Butanol ≥98.5%, GPR RECTAPUR®
: VWR Chemicals
Ethanol eurodenatured ≥80%, TechniSolv® (1:1:1)
: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Methanol, extra dry 99.9%, AcroSeal®
: Thermo Fisher Scientific