Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one,97% 1 * 0,25 g
Catalog # AMBDA100251-250MG
Supplier:
AMBEED
Expand image
4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one,97% 1 * 0,25 g
Catalog # AMBDA100251-250MG
Supplier:
AMBEED
Frequently Bought Together
2-Propanol ≥99.0% USP, Ph. Eur.
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethyl acetate, anhydrous (max. 0.005% H₂O) ≥99.8%
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
1,3-Propanediol ≥99.7%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Dichloromethane, anhydrous (max. 0.001% H₂O) ≥99.8% stabilised
Supplier: VWR Chemicals
Ethanol ≥96% (v/v), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Ethanolamine, Sigma-Aldrich®
Supplier: Merck