Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Cis-4-decen-1-ol,90+% 1 * 5 g
Catalog # AMBDA594932-5G
Supplier:
AMBEED
Expand image
Cis-4-decen-1-ol,90+% 1 * 5 g
Catalog # AMBDA594932-5G
Supplier:
AMBEED
Frequently Bought Together
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Residual solvents Class 2 - mix A (2013 rev) (16 components)
Supplier: Restek