Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1-tert-Butyl 2-methyl 3-bromo-1H-pyrrole-1,2-dicarboxylate,98% 1 * 0,1 g
Catalog # AMBDA711506-100MG
Supplier:
AMBEED
Expand image
1-tert-Butyl 2-methyl 3-bromo-1H-pyrrole-1,2-dicarboxylate,98% 1 * 0,1 g
Catalog # AMBDA711506-100MG
Supplier:
AMBEED
Frequently Bought Together
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific
Diethyl ether ≥99% stabilised for HPLC
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific