Order Entry
Slovenia
ContactUsLinkComponent
MiSeq System Instrument Configuration: RFID tracking for consumables MiSeq Control Software Local Run Manager Software Base Unit: Intel Core i7-2710QE 2.10 GHz CPU Memory: 16 GB RAM Hard Drive: 750 GB Operating System: Windows 7 embedded standard Te 1 * 1 ST
Catalog # IBSASY-410-1003
Supplier:  IBS Stock Items
MiSeq System Instrument Configuration: RFID tracking for consumables MiSeq Control Software Local Run Manager Software Base Unit: Intel Core i7-2710QE 2.10 GHz CPU Memory: 16 GB RAM Hard Drive: 750 GB Operating System: Windows 7 embedded standard Te 1 * 1 ST
Catalog # IBSASY-410-1003
Supplier:  IBS Stock Items
Frequently Bought Together