Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RECOMBINANT CLONEDETECT, ANTI-HUMAN IGG (Fc), FITC LABELED, STERILE, AZIDE-FREE 1 * 1 items
Catalog # MLDVK8495
Supplier:
Molecular Devices
Expand image
RECOMBINANT CLONEDETECT, ANTI-HUMAN IGG (Fc), FITC LABELED, STERILE, AZIDE-FREE 1 * 1 items
Catalog # MLDVK8495
Supplier:
Molecular Devices
Frequently Bought Together
VWR®, Breatheable Rayon Film Seals for Biological Cultures
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Supplier: VWR Collection
Serological pipettes
Supplier: Greiner Bio-One
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes with Dividers
Supplier: VWR Collection
VWR®, Serological Pipettes, Open End
Supplier: VWR Collection