Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Liver Meat Glucose Cysteine Broth (500G) 1 * 1 ST
Catalog # HIMEM1322-500G
Supplier:
HIMEDIA
Certificates
Expand image
Liver Meat Glucose Cysteine Broth (500G) 1 * 1 ST
Catalog # HIMEM1322-500G
Supplier:
HIMEDIA
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 5 L
Supplier: PanReac AppliChem
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Chloroform 99.8% stabilised for analysis
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals