PrecisION hASIC3 Recombinant Stable Cell Line 1 * 2 Vial
Catalog # DISXCYL3055
Supplier: DISCOVERX
Frequently Bought Together
Colour coders for cryovials, CryoColorCode, Nunc™
Supplier: Thermo Fisher Scientific
Assay microplates and blocks, 384-well, ScreenMates
Supplier: THERMO MATRIX TECHNOLOGIES
Chemical bottles, wide mouth, square
Supplier: RIXIUS
Micro packaging vials with thread, Nalgene®
Supplier: Thermo Fisher Scientific
AcroPrep™ 24-Well Receiver Plate
Supplier: Cytiva (Formerly Pall Lab)
Stoppers, press down, VacuCap®
Supplier: Simport Scientific
HTS TranswellRegistered Mark-96, 0.4 µm Pore Polyester Membrane Inserts 1 * 480 items
Supplier: STEMCELL Technologies
Cap inserts for cryogenic vials, TruCool®
Supplier: BIOCISION
Lids for microplates, Matrix
Supplier: THERMO MATRIX TECHNOLOGIES
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling
PCR plates, 96-well, Armadillo™
Supplier: Thermo Fisher Scientific
Filter plates, 96-well, AcroPrep™
Supplier: Cytiva (Formerly Pall Lab)