PathHunter U2OS IL1RL1/IL1RAP Dimerization Cell Line 1 * 2 Vial
Catalog # DISX93-1067C3
Supplier: DISCOVERX
PathHunter U2OS IL1RL1/IL1RAP Dimerization Cell Line 1 * 2 Vial
Catalog # DISX93-1067C3
Supplier: DISCOVERX
Frequently Bought Together
VWR®, Test Tubes without Rim
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR®, Spreaders, L-Shaped, Standard Line
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
Media bottles, square, sterile, Nalgene®
Supplier: Thermo Fisher Scientific
Test tubes
Supplier: Greiner Bio-One
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Centrifuge tubes, 15 ml, Corning®
Supplier: Corning
Petri dishes, square
Supplier: Greiner Bio-One
Multiple well cell culture plates, Costar®
Supplier: Corning
VWR®, Sampling Spoons
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Multiwell cell culture plates, Falcon®
Supplier: Corning