INTEGRATED STEAM GENERATOR (FOR UPTO 200L ACS) 1 * 1 items
PRIOPC/SGI/000
Frequently Bought Together
VWR®, Probes for Precise PT1000 Thermometer TD23
: VWR Collection
TRAY - SGLE WFR 150MM H22-60-0615 1 * 100 ST
: ENTEGRIS
Repetitive pipette, Multipette® E3 / E3x
: EPPENDORF
Stainless Steel Solid Modular Shelving Unit
: TEKNOMEK
Additional Top and Bottom Shelves
: Palbam
Heating baths, HB eco
: IKA
Single channel pipettes, variable volume
: EDVOTEK
A114 CAP SCREW M8x1 1 * 1 items
: TUTTNAUER
FRIDGE FORCED AIR 327L 1 * 1 items
: Liebherr