3-CHLORO-4-FLUORO-7,8-DIHYDRO-6H-ISOQUINOLIN-5-ONE 1 * 10 g
Catalog # PHBLPBLID0506-10G
Supplier: Pharmablock (USA)
3-CHLORO-4-FLUORO-7,8-DIHYDRO-6H-ISOQUINOLIN-5-ONE 1 * 10 g
Catalog # PHBLPBLID0506-10G
Supplier: Pharmablock (USA)
Frequently Bought Together
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.8%
Supplier: VWR Chemicals
1,3-Dioxolane ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Tetrahydrofuran, anhydrous (max. 0.003% H₂O) ≥99.9% stabilised
Supplier: VWR Chemicals
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Cyclohexanone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Hydrogen peroxide 30%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
tert-Butanol, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% for LC-MS
Supplier: Thermo Fisher Scientific
Ethylbenzene 99.8%, pure
Supplier: Thermo Fisher Scientific